The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201644.2 | NP_964006.2 | 1 - 1 | 553 | 93 |
GenBank mRNA | AY227200.1 | - | 1 - 1 | 532 | 93 |
BC019434.1 | - | 1 - 1 | 527 | 93 | |
BC138699.1 | - | 1 - 1 | 532 | 93 | |
BC146021.1 | - | 1 - 1 | 532 | 93 |