Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- 1700065F09Rik
- Entrez Gene ID
-
78463
- Gene Name
- RIKEN cDNA 1700065F09 gene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK018878.1 |
- |
1 - 1 |
151 |
103 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY706880.1 |
- |
1 |
150 |
|
Chr. 2 - 85086620 - 85086981 [+] on Build GRCm38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 4930443O20 gene |
4930443O20Rik |
|
-159.0
|
Anti-sense |
Chr. - 85073614 - 85086461 on Build GRCm38 |
Downstream |
apelin receptor |
Aplnr |
APJ, Agtrl1, msr/apj |
49.4kb
|
Sense |
Chr. - 85136360 - 85139923 on Build GRCm38 |
View Full Product Details