Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gm31452
- Entrez Gene ID
-
102633689
- Gene Name
- predicted gene, 31452
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 107063405 - 107148551 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK172672.1 |
- |
3 - 4 |
512 |
94 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY766092.1 |
- |
1 - 2 |
361 |
|
Chr. 13 - 107085990 - 107087797 [+] on Build GRCm38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 3830408C21 gene |
3830408C21Rik |
|
-52.5kb
|
Sense |
Chr. - 107022570 - 107033518 on Build GRCm38 |
Downstream |
apolipoprotein O, pseudogene |
Apoo-ps |
Apoo, EG621156 |
326.4kb
|
Anti-sense |
Chr. - 107414163 - 107414767 on Build GRCm38 |
View Full Product Details