Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r77
Entrez Gene ID
628946
Gene Name
vomeronasal 1 receptor 77
Gene Aliases
Gm6935
Chromosome Location
Chr.7: 12036846 - 12042219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166731.1 NP_001160203.1 1 - 1 752 102

View Full Product Details