Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Defb25
Entrez Gene ID
654459
Gene Name
defensin beta 25
Gene Aliases
BD-25, OTTMUSG00000015862, mBD-25
Chromosome Location
Chr.2: 152622356 - 152623053 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039122.1 NP_001034211.1 1 - 2 88 94
GenBank mRNA BC132662.1 - 1 - 2 84 94
DQ012036.1 - 1 - 2 88 94

View Full Product Details