The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077362.2 | NP_001070830.1 | 5 - 6 | 821 | 113 |
NM_001287800.1 | NP_001274729.1 | 5 - 6 | 765 | 113 | |
NM_010211.3 | NP_034341.2 | 6 - 7 | 874 | 113 | |
GenBank mRNA | AF114380.1 | - | 4 - 5 | 735 | 113 |
AK128904.1 | - | 6 - 7 | 848 | 113 | |
BC029024.1 | - | 5 - 6 | 766 | 113 | |
BC031120.1 | - | 4 - 5 | 715 | 113 | |
U41739.1 | - | 5 - 6 | 765 | 113 | |
U77039.1 | - | 6 - 7 | 818 | 113 |