The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008463.2 | NP_032489.1 | 1 - 2 | 82 | 85 |
GenBank mRNA | AF247643.1 | - | 1 - 2 | 51 | 85 |
AF419251.1 | - | - | 215 | 85 | |
BC120550.1 | - | 1 - 2 | 40 | 85 | |
BC137708.1 | - | 1 - 2 | 40 | 85 | |
U10091.1 | - | 1 - 2 | 82 | 85 |