Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101106475
Entrez Gene ID
101106475
Gene Name
RNA-binding protein 14
Gene Aliases
RBM14
Chromosome Location
Chr.21: 46424832 - 46440803 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959711.1 XP_027815512.1 2 - 3 1912 87

View Full Product Details