Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL12
Entrez Gene ID
101110231
Gene Name
F-box and leucine rich repeat protein 12
Gene Aliases
-
Chromosome Location
Chr.5: 12685345 - 12692211 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008505.3 XP_004008554.1 1 - 2 402 90

View Full Product Details