Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCAT
Entrez Gene ID
100302343
Gene Name
lecithin-cholesterol acyltransferase
Gene Aliases
-
Chromosome Location
Chr.14: 38196268 - 38199517 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162568.1 NP_001156040.1 1 - 2 149 72
GenBank mRNA FJ958346.1 - 1 - 2 149 72

View Full Product Details