Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIX4
Entrez Gene ID
101104155
Gene Name
SIX homeobox 4
Gene Aliases
-
Chromosome Location
Chr.7: 75731241 - 75743594 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971911.1 XP_027827712.1 2 - 3 1851 72

View Full Product Details