The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199068.1 | NP_001185997.1 | 3 - 4 | 349 | 93 |
GenBank mRNA | HQ433587.1 | - | 3 - 4 | 349 | 93 |