Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HYAL2
Entrez Gene ID
443154
Gene Name
hyaluronidase 2
Gene Aliases
oHyal2
Chromosome Location
Chr.19: 52008120 - 52013227 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009754.1 NP_001009754.1 2 - 3 1055 90
XM_012099237.3 XP_011954627.1 3 - 4 1182 90
XM_012099238.3 XP_011954628.1 3 - 4 1181 90
GenBank mRNA AF411974.1 - 2 - 3 1055 90
JX231101.1 - 2 - 3 1023 90

View Full Product Details