Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNLT3
Entrez Gene ID
443256
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
TCTE1L
Chromosome Location
Chr.X: 38783883 - 38794776 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009762.1 NP_001009762.1 3 - 4 208 89
XM_027962770.1 XP_027818571.1 3 - 4 230 89
GenBank mRNA AY205338.1 - 3 - 4 208 89

View Full Product Details