Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIB4
Entrez Gene ID
100303602
Gene Name
calcium and integrin binding family member 4
Gene Aliases
-
Chromosome Location
Chr.3: 35266879 - 35319466 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164713.1 NP_001158185.1 3 - 4 186 94
GenBank mRNA FJ039532.1 - 3 - 4 186 94
KJ425421.1 - 3 - 4 185 94

View Full Product Details