Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COTL1
Entrez Gene ID
101103415
Gene Name
coactosin like F-actin binding protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 11448786 - 11493210 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014943.2 XP_004014992.1 1 - 2 306 80

View Full Product Details