Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJB1
Entrez Gene ID
101122104
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
Gene Aliases
-
Chromosome Location
Chr.5: 9600073 - 9611995 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008469.4 XP_004008518.1 2 - 3 886 82
XM_027969483.1 XP_027825284.1 2 - 3 949 82

View Full Product Details