Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LXN
Entrez Gene ID
101108098
Gene Name
latexin
Gene Aliases
-
Chromosome Location
Chr.1: 249571935 - 249579601 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003214.4 XP_004003263.2 3 - 4 638 69

View Full Product Details