Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFSD9
Entrez Gene ID
101108476
Gene Name
major facilitator superfamily domain containing 9
Gene Aliases
-
Chromosome Location
Chr.3: 105491284 - 105505412 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007326.4 XP_004007375.2 4 - 5 419 63

View Full Product Details