Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCM2
Entrez Gene ID
101105538
Gene Name
glial cells missing transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.20: 48844576 - 48850976 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019134.4 XP_004019183.1 3 - 4 459 78

View Full Product Details