Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC52
Entrez Gene ID
101112100
Gene Name
leucine rich repeat containing 52
Gene Aliases
-
Chromosome Location
Chr.1: 125015809 - 125037215 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002720.4 XP_004002769.1 1 - 2 657 99

View Full Product Details