Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA10
Entrez Gene ID
101108240
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
-
Chromosome Location
Chr.11: 1940193 - 1991239 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974001.1 XP_027829802.1 7 - 8 1159 61

View Full Product Details