Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FADS6
Entrez Gene ID
101102657
Gene Name
fatty acid desaturase 6
Gene Aliases
-
Chromosome Location
Chr.11: 6745811 - 6761140 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974049.1 XP_027829850.1 2 - 3 371 69

View Full Product Details