Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH4A
Entrez Gene ID
101105003
Gene Name
radial spoke head component 4A
Gene Aliases
-
Chromosome Location
Chr.8: 23311446 - 23326173 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011178.4 XP_004011227.3 4 - 5 1919 64

View Full Product Details