Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK35
Entrez Gene ID
101114546
Gene Name
serine/threonine kinase 35
Gene Aliases
-
Chromosome Location
Chr.13: 55303323 - 55344978 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976886.1 XP_027832687.1 2 - 3 905 98

View Full Product Details