Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5MG
Entrez Gene ID
101123491
Gene Name
ATP synthase membrane subunit g
Gene Aliases
ATP5L
Chromosome Location
Chr.15: 31902441 - 31910936 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979248.1 XP_027835049.1 2 - 3 484 69

View Full Product Details