Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMAC2
Entrez Gene ID
101122972
Gene Name
distal membrane arm assembly complex 2
Gene Aliases
ATP5SL
Chromosome Location
Chr.14: 53798974 - 53804749 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015271.3 XP_004015320.2 5 - 6 636 101

View Full Product Details