Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CHSY3
Entrez Gene ID
101104485
Gene Name
chondroitin sulfate synthase 3
Gene Aliases
-
Chromosome Location
Chr.5: 22649349 - 22939941 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969960.1 XP_027825761.1 1 - 2 868 102

View Full Product Details