Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALML6
Entrez Gene ID
101123572
Gene Name
calmodulin like 6
Gene Aliases
-
Chromosome Location
Chr.12: 54586229 - 54608122 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976078.1 XP_027831879.1 5 - 6 842 86

View Full Product Details