Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRX1
Entrez Gene ID
101103742
Gene Name
paired related homeobox 1
Gene Aliases
-
Chromosome Location
Chr.12: 41509667 - 41588745 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099201.2 XP_014954687.2 1 - 2 1341 135
XM_027975594.1 XP_027831395.1 1 - 2 1348 135

View Full Product Details