Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXRA5
Entrez Gene ID
101116543
Gene Name
matrix remodeling associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 1484361 - 1513583 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963681.1 XP_027819482.1 6 - 7 6838 86
XM_027963682.1 XP_027819483.1 6 - 7 6698 86

View Full Product Details