Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGF29
Entrez Gene ID
101113816
Gene Name
SAGA complex associated factor 29
Gene Aliases
CCDC101
Chromosome Location
Chr.24: 26626009 - 26658266 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012104338.3 XP_011959728.1 5 - 6 460 99
XM_012104339.3 XP_011959729.1 6 - 7 444 99

View Full Product Details