Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PJVK
Entrez Gene ID
101102040
Gene Name
pejvakin
Gene Aliases
DFNB59
Chromosome Location
Chr.2: 142282982 - 142289288 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004545.4 XP_004004594.1 1 - 2 211 128

View Full Product Details