Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBDS
Entrez Gene ID
101111067
Gene Name
SBDS ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.24: 29088775 - 29094521 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020948.4 XP_004020997.1 2 - 3 513 93

View Full Product Details