Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG4
Entrez Gene ID
101118503
Gene Name
BAG cochaperone 4
Gene Aliases
-
Chromosome Location
Chr.26: 36331524 - 36362092 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104605.2 XP_014960091.2 3 - 4 668 102

View Full Product Details