Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
AGO2
Entrez Gene ID
100009457
Gene Name
argonaute RISC catalytic component 2
Gene Aliases
EIF2C2
Chromosome Location
Chr.19: 11598574 - 11599578 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082710.1 NP_001076179.1 - 1340 81
GenBank mRNA AF005355.1 - - 1340 81

View Full Product Details