Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNJ11
Entrez Gene ID
100008648
Gene Name
potassium inwardly rectifying channel subfamily J member 11
Gene Aliases
KIR6.2
Chromosome Location
Chr.Unknown: 1405335 - 1406511 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082017.1 NP_001075486.1 - 950 87
GenBank mRNA AF006262.1 - - 950 87
AF455118.1 - - 87
AJ309932.1 - - 87
U92804.1 - - 87

View Full Product Details