Gene Transcripts

Gene Symbol
Fh
Entrez Gene ID
24368
Gene Name
fumarate hydratase
Gene Aliases
Fh1
Chromosome Location
Chr. 13: 93651486 - 93677371 on Build Rnor_6.0
UniGene ID
Rn.29782
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017005.2 NP_058701.2 3, 4 383
GenBank mRNA BC087598.1 - 3, 4 383
J04473.1 - 3, 4 394

View Full Product Details