Gene Transcripts

Gene Symbol
Rbm31y
Entrez Gene ID
302780
Gene Name
RNA binding motif 31, Y-linked
Gene Aliases
RGD1562219
Chromosome Location
Chr. X: 98093734 - 98095719 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_229073.3 XP_229073.1 1 227

View Full Product Details