Gene Transcripts

Gene Symbol
Capn12
Entrez Gene ID
308476
Gene Name
calpain 12
Gene Aliases
Chromosome Location
Chr. 1: 87066289 - 87078348 on Build Rnor_6.0
UniGene ID
Rn.219171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110808.1 NP_001104278.1 5 749

View Full Product Details