Gene Transcripts

Gene Symbol
Fastkd1
Entrez Gene ID
311112
Gene Name
FAST kinase domains 1
Gene Aliases
RGD1563531
Chromosome Location
Chr. 3: 55925598 - 55951584 on Build Rnor_6.0
UniGene ID
Rn.226110
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191738.1 NP_001178667.1 7 2392
XM_006234311.1 XP_006234373.1 7 1201

View Full Product Details