Gene Transcripts

Gene Symbol
Fbxo15
Entrez Gene ID
361354
Gene Name
F-box protein 15
Gene Aliases
Chromosome Location
Chr. 18: 81785408 - 81882796 on Build Rnor_6.0
UniGene ID
Rn.24143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108436.1 NP_001101906.1 5 708

View Full Product Details