Gene Transcripts

Gene Symbol
Krt36
Entrez Gene ID
287698
Gene Name
keratin 36, type I
Gene Aliases
Ka31, Krt1-5
Chromosome Location
Chr. 10: 88016602 - 88019967 on Build Rnor_6.0
UniGene ID
Rn.2873
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008759.1 NP_001008759.1 3, 4 689

View Full Product Details