Gene Transcripts

Gene Symbol
Aox2
Entrez Gene ID
316421
Gene Name
aldehyde oxidase 2
Gene Aliases
Aox2p, Aox3l1
Chromosome Location
Chr. 9: 65175452 - 65266496 on Build Rnor_6.0
UniGene ID
Rn.215801
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008522.1 NP_001008522.1 1 28
XM_008767106.1 XP_008765328.1 2 1657
GenBank mRNA AY665588.1 - 1 28

View Full Product Details