Gene Transcripts

Gene Symbol
Vom1r3
Entrez Gene ID
494237
Gene Name
vomeronasal 1 receptor 3
Gene Aliases
V1re25
Chromosome Location
Chr. 1: 86429262 - 86430158 on Build Rnor_6.0
UniGene ID
Rn.129462
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009517.1 NP_001009517.1 1 53
GenBank mRNA AY510342.1 - 1 53

View Full Product Details