Gene Transcripts

Gene Symbol
Syce1
Entrez Gene ID
502372
Gene Name
synaptonemal complex central element protein 1
Gene Aliases
Chromosome Location
Chr. 1: 213522054 - 213534641 on Build Rnor_6.0
UniGene ID
Rn.159914
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025058.2 NP_001020229.2 3 202
XM_006230572.2 XP_006230634.1 3 119
GenBank mRNA BC083907.1 - 3 184
CK596157.1 - 3 202

View Full Product Details