Gene Transcripts

Gene Symbol
Asap3
Entrez Gene ID
684122
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
Gene Aliases
Chromosome Location
Chr. 5: 154552016 - 154596125 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764321.1 XP_008762543.1 17 1862

View Full Product Details