Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134763.1 | NP_001128235.1 | 6 | 491 | |
NM_031234.1 | NP_112513.1 | 6 | 491 | ||
XM_008769266.1 | XP_008767488.1 | 6 | 580 | ||
XM_008769267.1 | XP_008767489.1 | 6 | 580 | ||
XM_008769268.1 | XP_008767490.1 | 6 | 2630 | ||
GenBank mRNA | AB005052.1 | - | 6 | 491 | |
BC169096.1 | - | 6 | 478 | 7936362 |