Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM219A
Entrez Gene ID
700651
Gene Name
family with sequence similarity 219 member A
Gene Aliases
C15H9orf25, C9orf25
Chromosome Location
Chr.15: 49741550 - 49801019 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193839.2 NP_001180768.1 - 368 71
XM_002800103.3 XP_002800149.1 - 403 71
XM_015117356.2 XP_014972842.1 - 403 71
XM_015117357.2 XP_014972843.1 - 403 71
GenBank mRNA JU322134.1 - - 372 71
JU322135.1 - - 372 71
JU322219.1 - - 372 71
JU472014.1 - - 355 71
JV601564.1 - - 372 71

View Full Product Details