Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRTC2
Entrez Gene ID
715908
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
EGK_01342
Chromosome Location
Chr.1: 97103264 - 97114446 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261373.1 NP_001248302.1 - 1084 89
XM_015152564.2 XP_015008050.1 - 1161 89
XM_015152571.2 XP_015008057.1 - 1092 89
XM_015152577.2 XP_015008063.1 - 1063 89
XM_015152587.2 XP_015008073.1 - 1048 89
XM_015152597.2 XP_015008083.1 - 1423 89
XM_028845384.1 XP_028701217.1 - 1319 89
XM_028845386.1 XP_028701219.1 - 883 89
XM_028845391.1 XP_028701224.1 - 1304 89
GenBank mRNA JU333300.1 - - 1040 89
JU474613.1 - - 1081 89
JV046393.1 - - 1084 89

View Full Product Details